missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PATL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548187
This item is not returnable.
View return policy
Description
PATL2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
RNA-binding protein that acts as a translational repressor.
Specifications
| PATL2 | |
| Polyclonal | |
| Unconjugated | |
| PATL2 | |
| 4930424G05Rik; hPat1a; PAT1 homolog 2; Pat1a; PAT1-like protein 2; Patl2; protein associated with topoisomerase II homolog 2; protein associated with topoisomerase II homolog 2 (yeast); protein PAT1 homolog 2; Protein PAT1 homolog a | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 197135 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| C9JE40 | |
| PATL2 | |
| KLH conjugated synthetic peptide between 168-196 amino acids from the Central region of human PATL2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction