missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress IL4R/IL2RG Dimerization Assay
Description
The PathHunter™ IL4R/IL2RG Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. IL-4) induced dimerization of IL4R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL4R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL4R/IL2RG , detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | IL4R/IL2RG Dimerization |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | IL-4 |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | IL4R/IL2RG |
| Cell Line | U2OS |
| Target | IL4R/IL2RG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?