missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress EGFR/EGFR Dimerization Assay

Product Code. 17314169
Encompass
Change view
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17314169 1,000 dp (10 x 96-well) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17314169 Supplier DiscoverX Supplier No. 930989E3CP5L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress EGFR/EGFR Dimerization Assay measures ligand-induced dimerization of EGFR/EGFR . eXpress kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ EGFR/EGFR Dimerization eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This kit assesses ligand (e.g. EGF) induced dimerization of EGFR/EGFR receptors. This product uses EFC technology. The cell line included overexpresses EGFR target protein fused to PK alongside EGFR target protein fused to EA. Upon ligand-induced dimerization of EGFR/EGFR , detectable EFC signal is produced. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry EGFR/EGFR Dimerization
For Use With (Application) Assay-Ready eXpress Kits
Primary Agonist Recombinant Human EGF
Quantity 1,000 dp (10 x 96-well)
Target Entry EGFR/EGFR
Cell Line U2OS
Target EGFR/EGFR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.