Description
- Path-Chek Hygiene Test
- Rapid test system designed for the detection of a range of bacteria from work surfaces and the processing environment.
- Simple and instant swab test for the assessment of cleaning efficiency by the detection of protein and possible allergens.
Specifications
Specifications
| Detects | Protein, possible allergens |
| Test Type | Swab Test |
| Quantity | 100 Tests |
Product Title
Haben Sie Verbesserungsvorschläge?