Learn More
Novacyt - Microgen Bioproducts Path-Chek Coliforms Swab Test
Shop All Novacyt Group Products
Click to view available options
:
Detects: Coliforms, 100 tests
Description
- Path-Chek Hygiene Test
- Rapid test system designed for the detection of a range of bacteria from work surfaces and the processing environment.
- Simple and instant swab test for the detection of Coliforms by direct testing of surfaces.
Specifications
Specifications
| Description | Swab test, 100 tTests |
| For Use With (Application) | Coliform detection |