Learn More
Thermo Scientific™ Pasteurella Selective Agar
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
Description
This specialized agar medium is meticulously formulated to support the growth of Pasteurella species while inhibiting non-target organisms, providing accurate and reliable results for your microbiological testing needs.
Specifications
Specifications
| Color | Transparent |
| Content And Storage | 2°C to 12°C |
| Format | 90 mm |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Volume | 17 mL |
| Packaging Type | 1 x 10 Plates |
| Quantity | 10 Plates |