Learn More
KARTELL Pasteur Pipette
Product Code. 17838085
Change view
Click to view available options
Volume (Metric):
3.5 mL
4 mL
4.4 mL
5 mL
5.4 mL
7 mL
Sterility:
Non-sterile
Sterile
Drops/mL:
21
23
28
Unit Size:
Pack of 1000
Pack of 500
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Drops/mL Unit Size
17838085 7 mL Sterile 21 Pack of 500
17848085 4 mL Sterile 28 Pack of 500
10274991 4.4 mL Sterile - Pack of 1000
17818085 3.5 mL Sterile 23 Pack of 500
10275231 5.4 mL Sterile - Pack of 1000
17808085 4 mL Non-sterile 28 Pack of 500
17828085 5 mL Non-sterile 23 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17838085 Supplier KARTELL Supplier No. 00334/1
  • ONLINE EXCLUSIVE PRICE
    760.00 NOK
    680.17 NOK / Pack of 500
    Save 79.83 NOK
    10% Off
Estimated Shipment: 22-12-2026
These disposable Pasteur Pipettes are ideal for the precise transfer of samples.
  • Disposable PE pipette with bulb and graduations.
  • The slender tip guarantees precision transfer of the sample.
  • Sterile pipettes individually wrapped.
Specifications
Bulb Draw (Metric) 3.5 mL
Material Polyethylene
Sterility Sterile
Drops/mL 21
Graduated Yes
Graduations 3 mL/ Subgrad: 0.5 mL
Color Clear
Volume (Metric) 7 mL
Length (Metric) 150 mm
Type Pasteur Pipet
Disposable Single-use
Fit for Purpose Sterile