Learn More
Pasireotide ditrifluoroacetate, MedChemExpress
Description
Pasireotide (SOM230) ditrifluoroacetate, a long-acting cyclohexapeptide somatostatin analogue, can improve agonist activity at somatostatin receptors (subtypes sst1/2/3/4/5, pKi=8.2/9.0/9.1/<7.0/9.9, respectively). Pasireotide ditrifluoroacetate exhibits antisecretory, antiproliferative, and proapoptotic activity.
Specifications
Specifications
| Chemical Name or Material | Pasireotide ditrifluoroacetate |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C62H68F6N10O13 |
| Quantity | 1 mg |
| Synonym | SOM230 ditrifluoroacetate Pasireotide TFA salt |
| Solubility Information | DMSO : 100 mg/mL (78.42 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (26.14 mM; Need ultrasonic) |
| SMILES | O=C(O)C(F)(F)F.O=C(O)C(F)(F)F.NCCCC[C@@H](C(N[C@H]1CC2=CC=C(C=C2)OCC3=CC=CC=C3)=O)NC([C@H](NC([C@H](C4=CC=CC=C4)NC([C@H]5N(C([C@H](CC6=CC=CC=C6)NC1=O)=O)C[C@H](OC(NCCN)=O)C5)=O)=O)CC7=CNC8=C7C=CC=C8)=O |
| Molecular Weight (g/mol) | 1275.25 |
| Formula Weight | 1275.25 |
| Show More |