Learn More
Heathrow Scientific™ Partitions for Cryogenic Vial BVoxes
GREENER_CHOICE
Product Code. 12355043 Shop All Thermo Scientific CO₂ Incubators Products
Product Code. 12355043
Brand: Heathrow Scientific™ HS2860E
Match the partition to your tube requirements. Heathrow Scientific™ Partitions for Cryogenic Vial Boxes stabilize 1.5/2.0mL tubes inside their boxes during storage and transport.
  • Keeps vials upright during storage and transport
  • For use with 1.5/2.0mL tubes
Specifications
Material Cardboard
Product Type Box Divider Insert
Holds 100 x 1.5/2.0mL tubes
Color White
For Use With (Equipment) Cryogenic vial boxes
Width (Metric) 13.3cm
Length (Metric) 13.3cm