missing translation for 'onlineSavingsMsg'
Learn More

PARP9 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Product Code. 16847444 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16847444 150 μL 150µL
16857444 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16847444

Brand: Proteintech 175351AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

In concert with DTX3L plays a role in PARP1-dependent DNA damage repair. PARP1-dependent PARP9/BAL1-DTX3L-mediated ubiquitination promotes the rapid and specific recruitment of 53BP1/TP53BP1, UIMC1/RAP80, and BRCA1 to DNA damage sites. Involved in inducing the expression of IFN-gamma-responsive genes.

Specifications

Antigen PARP9
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PARP9
Gene Accession No. Q8IXQ6
Gene Alias B aggressive lymphoma protein, BAL, BAL1, PARP 9, PARP9, Poly [ADP ribose] polymerase 9
Gene Symbols PARP9
Host Species Rabbit
Immunogen PARP9 Fusion Protein Ag11587
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303905, 83666
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.