missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP9 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17535-1-AP-20UL
This item is not returnable.
View return policy
Description
In concert with DTX3L plays a role in PARP1-dependent DNA damage repair. PARP1-dependent PARP9/BAL1-DTX3L-mediated ubiquitination promotes the rapid and specific recruitment of 53BP1/TP53BP1, UIMC1/RAP80, and BRCA1 to DNA damage sites. Involved in inducing the expression of IFN-gamma-responsive genes.Specifications
| PARP9 | |
| Polyclonal | |
| Unconjugated | |
| PARP9 | |
| B aggressive lymphoma protein, BAL, BAL1, PARP 9, PARP9, Poly [ADP ribose] polymerase 9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 303905, 83666 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IXQ6 | |
| PARP9 | |
| PARP9 Fusion Protein Ag11587 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction