Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Threaded Adapter Elbow
Product Code. 11719739
Click to view available options
Tubing Size:
1/2 in.
1/4 in.
1/8 in.
3/8 in.
Unit Size:
Each
Product Code. 11719739
Brand: Cole-Parmer™ 3132014
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
  • Superior chemical resistance with durable PFA composition.
  • Get a tight seal at higher pressures.
  • Withstand higher pressures than barbed fittings.
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Description Compression x NPT(M) Connection
Autoclavable Autoclavable
Material PFA
Tubing Size 1/8 in.
Outlet Port 1/8 x 1/8 in.
Sterilization Method Autoclave or Ethylene Oxide
Thread Size 1/8 in. NPT(M)
Color Clear
Product Type Compression Threaded Adapter Elbow