Learn More
Cole-Parmer™ Parker Hannifin™ PFA Compression Female Threaded Adapter
Excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings
610.00 NOK - 945.00 NOK
Specifications
Product Type | Compression Threaded Adapter |
---|---|
Description | Compression x NPT(F) |
Autoclavable | Autoclavable |
Material | PFA |
Sterilization Method | Autoclave or Ethylene Oxide |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11785489
|
Cole-Parmer™
31320-52 |
1/4 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11769729
|
Cole-Parmer™
31320-72 |
1/2 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11770129
|
Cole-Parmer™
31320-51 |
1/8 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Beschreibung
- Superior chemical resistance with durable PFA composition.
- Get a tight seal at higher pressures.
- Withstand higher pressures than barbed fittings.
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Spezifikation
Compression Threaded Adapter | |
Autoclavable | |
Autoclave or Ethylene Oxide |
Compression x NPT(F) | |
PFA | |
Clear |