missing translation for 'onlineSavingsMsg'
Learn More

Parimifasor, MedChemExpress

Product Code. p-200098728
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476997 5 mg 5mg
16470607 50 mg 50mg
16465206 100 mg 100mg
16454278 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476997 Supplier Medchem Express Supplier No. HY109098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Parimifasor (LYC30937) is an immunomodulator, with anti-inflammatory activity.

Parimifasor (LYC30937) is an immunomodulator, with anti-inflammatory activity.

Specifications

Chemical Name or Material Parimifasor
CAS 1796641-10-5
Color White
Purity Grade Notes Research
Molecular Formula C18H11Cl2F4N5O
Quantity 5 mg
Synonym LYC30937
Solubility Information DMSO : 66.67 mg/mL (144.87 mM; Need ultrasonic)
SMILES O=C(C1=CC(Cl)=CC=C1)N/C(NC2=NNC(C(F)(F)F)=C2)=N/C3=CC(Cl)=CC(F)=C3
Molecular Weight (g/mol) 460.21
Formula Weight 460.21
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.