Learn More
Parimifasor, MedChemExpress
Product Code. 16476997
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476997 5 mg 5mg
16470607 50 mg 50mg
16465206 100 mg 100mg
16454278 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16476997 Supplier Medchem Express Supplier No. HY109098
MedChemExpress Parimifasor (LYC30937) is an immunomodulator, with anti-inflammatory activity.
Parimifasor (LYC30937) is an immunomodulator, with anti-inflammatory activity.
Specifications
Chemical Name or Material Parimifasor
CAS 1796641-10-5
Color White
Purity Grade Notes Research
Molecular Formula C18H11Cl2F4N5O
Quantity 5 mg
Synonym LYC30937
Solubility Information DMSO : 66.67 mg/mL (144.87 mM; Need ultrasonic)
SMILES O=C(C1=CC(Cl)=CC=C1)N/C(NC2=NNC(C(F)(F)F)=C2)=N/C3=CC(Cl)=CC(F)=C3
Molecular Weight (g/mol) 460.21
Formula Weight 460.21
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation