Learn More
Pargyline hydrochloride, MedChemExpress
Product Code. 16411986
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. 16411986
Brand: Medchem Express HYA0091
MedChemExpress Pargyline hydrochloride is an irreversible monoamine oxidase (MAO) inhibitor with Kis of 13 μM and 0.5 μM for MAO-A and MAO-B, respectively. Pargyline hydrochloride has antihypertensive and anticancer activities.
Pargyline hydrochloride is an irreversible monoamine oxidase (MAO) inhibitor with Kis of 13 μM and 0.5 μM for MAO-A and MAO-B, respectively. Pargyline hydrochloride has antihypertensive and anticancer activities.
Specifications
Chemical Name or Material Pargyline hydrochloride
Physical Form Solid
CAS 306-07-0
Color Off-White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C11H14ClN
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 100 mg/mL (511.01 mM; Need ultrasonic) ∣DMSO : ≥ 32 mg/mL (163.52 mM)
SMILES C#CCN(C)CC1=CC=CC=C1.[H]Cl
Molecular Weight (g/mol) 195.69
Formula Weight 195.69
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.