Learn More
Paraxanthine-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Paraxanthine 7-methyl-d3 |
| CAS | 65566-70-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7 2H3 H5 N4 O2 |
| Quantity | 10 mg |
| Synonym | 1H-Purine-2,6-dione','3,7-dihydro-1-methyl-7-(methyl-d3)-, Paraxanthine-7-(methyl-d3) |
| InChI Formula | InChI=1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)11(2)7(13)9-5/h3H,1-2H3,(H,9,13)/i1D3 |
| SMILES | [2H]C([2H])([2H])n1cnc2NC(=O)N(C)C(=O)c12 |
| IUPAC Name | 1-methyl-7-(trideuteriomethyl)-3H-purine-2,6-dione |
| Molecular Weight (g/mol) | 183.18 |
| Show More |