missing translation for 'onlineSavingsMsg'
Learn More

Pararosaniline hydrochloride, 90+%, Thermo Scientific Chemicals

Product Code. 15451457
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451457 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15451457 Supplier Thermo Scientific Alfa Aesar Supplier No. 036540.18
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

pH indicator 1.2 (purple) to 3.0 (red)

Pararosaniline hydrochloride is used to dye polyacrylonitrile fibers. It is also is used as a colorimetric test for aldehydes, in the Schiff test. It is the only basic fuchsine component suitable for making the aldehyde-fuchsine stain for pancreatic islet beta cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.824
MDL Number MFCD00001657
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
Synonym basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin
PubChem CID 11292
ChEBI CHEBI:87663
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl

Specifications

Melting Point 268°C to 270°C (decomposition)
Quantity 50 g
Beilstein 4164603
Solubility Information Soluble in water (2-3mg/ml),ethanol (2-25mg/ml),ether,and diethyl ether.
Formula Weight 323.83
Percent Purity ≥90%
Physical Form Powder
Chemical Name or Material Pararosaniline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.