Learn More
Paraffin, Supelco™
Product Code. 15877540
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877540 25 L 25 L
15854930 1 L 1 L
15867540 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15877540 Supplier Supelco Supplier No. 1071609026
 Restricted Item
Estimated Shipment: 04-02-2027
Viscous Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 8042-47-5
Specifications
Boiling Point 35°C to 36°C
Product Type Tissue Embedding Media
Quantity 25 L
Form Solution