Learn More
Pappalysin-1/PAPP-A Antibody (10E2cc) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NB100-73058JF525
Product Code. 30171809
  • 4580.00 NOK / 0.20mL
Description
Pappalysin-1/PAPP-A Monoclonal antibody specifically detects Pappalysin-1/PAPP-A in Human samples. It is validated for ELISA,Immunoprecipitation,Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pappalysin-1/PAPP-A
Monoclonal
Janelia Fluor 525
ASBABP2, DIPLA1, EC 3.4.24.79, IGFBP-4aseaspecific BCL2 ARE-binding protein 2, IGF-dependent IGFBP-4 protease, insulin-like growth factor-dependent IGF binding protein-4 protease, Insulin-like growth factor-dependent IGF-binding protein 4 protease, PAPA, PAPPA1, PAPP-Adifferentially placenta 1 expressed protein, pappalysin-1, pregnacy-associated plasma protein A, Pregnancy-associated plasma protein A, pregnancy-associated plasma protein A, pappalysin 1
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified human PAPP-A/proMBP (heterotetrameric PAPP-A, htPAPP-A) complex
0.2 mL
Cancer
5069
Store at 4°C in the dark.
IgG2b
ELISA, Immunoprecipitation, Sandwich ELISA
10E2cc
50mM Sodium Borate
Mouse
Protein A purified