Learn More
PAPPA Mouse anti-Human, Unconjugated, Clone: 1G5A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66962-1-IG-20UL
This item is not returnable.
View return policy
Description
The PAPPA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PAPPA. This antibody recognizes human antigen. The PAPPA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PAPPA | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 1G5A7 | |
| Unconjugated | |
| 20 μL | |
| 5069 | |
| IgG1 |