Learn More
PAPPA Monoclonal Antibody (PAPP A 1-24)
GREENER_CHOICE
Product Code. 11554112
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
200µg
Product Code. Quantity unitSize
11554112 400 μL 200µg
1 options
Product Code. 11554112
Brand: Invitrogen ABS0062402
PAPPA Monoclonal Antibody (PAPP A 1-24)
TRUSTED_SUSTAINABILITY
Specifications
Antigen PAPPA
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone PAPP A 1-24
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene PAPPA
Gene Accession No. Q13219
Gene Alias 8430414N03Rik; ASBABP2; aspecific BCL2 ARE-binding protein 2; differentially placenta 1 expressed protein; DIPLA1; IGFBP-4 ase; IGFBP-4ase; IGF-dependent IGFBP-4 protease; insulin-like growth factor-dependent IGF binding protein-4 protease; insulin-like growth factor-dependent IGF-binding protein 4 protease; PAG1; PAPA; PAPP A; PAPPA; PAPP-A; PAPPA1; Pappalysin; pappalysin 1; pappalysin-1; pregnacy-associated plasma protein A; pregnancy-associated plasma protein A; pregnancy-associated plasma protein A, pappalysin 1; RP11-45A16.1
Gene Symbols PAPPA
Host Species Mouse
Immunogen Purified human PAPP-A.
Purification Method Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5069
Target Species Human
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ