missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAMR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55310-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to PAMR1.
PAMR1 may be involved in regeneration of skeletal muscle. Although related to peptidase S1 family, lacks the conserved active Ser residue in position 665 which is replaced by a Thr, suggesting that it has no protease activity.Specifications
PAMR1 | |
Polyclonal | |
Unconjugated | |
PAMR1 | |
DKFZP586H2123, FP938, Inactive serine protease PAMR1, PAMR1, RAMP, UNQ699/PRO1344 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
210622, 25891, 311252 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UXH9, Q8BU25 | |
Pamr1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |