missing translation for 'onlineSavingsMsg'
Learn More

PAM16 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7207208 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16854454 20 μL 20µL
16844454 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16854454

Brand: Proteintech 153211AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Regulates ATP-dependent protein translocation into the mitochondrial matrix. Inhibits DNAJC19 stimulation of HSPA9/Mortalin ATPase activity.

Specifications

Antigen PAM16
Applications Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PAM16
Gene Accession No. Q9Y3D7
Gene Alias CGI 136, Magmas, PAM16, TIM16, TIMM16
Gene Symbols PAM16
Host Species Rabbit
Immunogen PAM16 Fusion Protein Ag0970
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51025
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.