Learn More
Palonosetron-3-ene N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | Palonosetron-3-ene N-Oxide |
| CAS | 1021456-82-5 |
| Molecular Formula | C19 H22 N2 O2 |
| Quantity | 100 mg |
| Synonym | 1H-Benz[de]isoquinolin-1-one','2,4,5,6-tetrahydro-2-[(3S)-1-oxido-1-azabicyclo[2.2.2]oct-3-yl]-, 2,4,5,6-Tetrahydro-2-[(3S)-1-oxido-1-azabicyclo[2.2.2]oct-3-yl]-1H-benz[de]isoquinolin-1-one, Palonosetron-3-ene N-Oxide |
| InChI Formula | InChI=1S/C19H22N2O2/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-20(19)17-12-21(23)9-7-13(17)8-10-21/h2,4,6,11,13,17H,1,3,5,7-10,12H2/t13?,17-,21?/m1/s1 |
| SMILES | [O-][N+]12CCC(CC1)[C@@H](C2)N3C=C4CCCc5cccc(C3=O)c45 |
| IUPAC Name | 2-[(3S)-1-oxido-1-azoniabicyclo[2.2.2]octan-3-yl]-5,6-dihydro-4H-benzo[de]isoquinolin-1-one |
| Molecular Weight (g/mol) | 310.39 |
| Formula Weight | 310.1681 |