Learn More
Palmitoleic Acid, TRC
Product Code. 30136529
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136529 1 g 1g
30133742 250 mg 250mg
30004504 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136529 Supplier Toronto Research Chemicals Supplier No. TRCP1449001G
 Restricted Item
Estimated Shipment: 26-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 373-49-9
Molecular Formula C16 H30 O2
Molecular Weight (g/mol) 254.41
Synonym 9-Hexadecenoic acid, (9Z)- (9CI, ACI), (9Z)-9-Hexadecenoic acid (ACI), 9-Hexadecenoic acid, (Z)- (8CI), Palmitoleic acid (7CI), (Z)-9-Hexadecenoic acid, (Z)-Hexadec-9-enoic acid, 9-cis-Hexadecenoic acid, 9-Hexadecenoic acid, 9Z-Hexadecenoic acid, C16:1, cis-9-Hexadecenoic acid, cis-Palmitoleic acid, cis-δ9-Hexadecenoic acid, Oleopalmitic acid, Zoomeric acid
IUPAC Name (Z)-hexadec-9-enoic acid
SMILES CCCCCC\C=C/CCCCCCCC(=O)O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C16H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h7-8H,2-6,9-15H2,1H3,(H,17,18)/b8-7-
Formula Weight 254.2246
Percent Purity >95%
Chemical Name or Material Palmitoleic acid
Grade HPLC