missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid, Sigma-Aldrich™

Product Code. 15876240
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876240 1 kg 1kg
15818320 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15876240 Supplier Sigma Aldrich Supplier No. 8005081000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-10-3

Specifications

Melting Point 61°C to 63°C
Density 0.852 g/cm3 at 20°C
Boiling Point 271.4°C/133 hPa
Molecular Formula CH3(CH2)14COOH
MDL Number MFCD00002747
Quantity 1 kg
Synonym Palmitic acid, Hexadecanoic acid, Palmitic acid
Molecular Weight (g/mol) 256.42
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.