Learn More
Palladium Matrix Modifier, Supelco™
Product Code. 15814970
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814970 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15814970 Supplier Supelco Supplier No. 1072890050
 Restricted Item
Estimated Shipment: 01-12-2026
For graphite furnace AAS c(Pd) = 10.0 ± 0.2 g/l (Pd(NO3)2 / HNO3 ca. 15%)
TRUSTED_SUSTAINABILITY
Specifications
Density 1.11 g/cm3 at 20°C
Type Palladium Modifier
Quantity 50 mL
Grade Research
Chemical Name or Material Palladium Matrix Modifier