Learn More
Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, nominally 50% water, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
2 g
10 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Physical Form | Powder |
Quantity | 2 g |
Odor | Odorless |
Chemical Name or Material | Palladium hydroxide, Pd 5% on carbon powder, Type A403002-5, Nominally 50% water |