Learn More
Brady™ Paint
Product Code. p-8004350
Change view
Click to view available options
Color:
Black
White
Unit Size:
Each
Product Code. Color unitSize
11741106 Black Each
11731106 White Each
2 options
Product Code. 11741106
Brand: Brady™ 804319
Paint for floor and road markings.
  • For Floor Marking.
  • For Traffic Control.
Specifications
Quantity 1 Ea.
Alternate Part Number PEINTURE ROUTIERE (5KG) NOIR
Product Type Paint
For Use With (Application) Floor Marking, Traffic Control
Color Black