Learn More
Brady™ Paint
Description
- For Floor Marking.
- For Traffic Control.
Specifications
Specifications
| Quantity | 1 Ea. |
| Alternate Part Number | PAINT FOR ROADS WHITE 30 L |
| Product Type | Paint |
| For Use With (Application) | Floor Marking, Traffic Control |
| Color | White |