Learn More
PAFAH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10085-1-AP-20UL
Additional Details : Weight : 0.00010kg
Beschreibung
Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.Spezifikation
PAFAH2 | |
Polyclonal | |
Unconjugated | |
PAFAH2 | |
HSD PLA2, PAFAH2, SD PLA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100163, 313611, 5051 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P83006, Q8VDG7, Q99487 | |
PAFAH2 | |
PAFAH2 Fusion Protein Ag0124 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |