missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PADI4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17373-1-AP-150UL
This item is not returnable.
View return policy
Description
Catalyzes the citrullination/deimination of arginine residues of proteins. Citrullinates histone H3 at ′Arg-8′ and/or ′Arg-17′ and histone H4 at ′Arg-3′, which prevents their methylation by CARM1 and HRMT1L2/PRMT1 and represses transcription. Citrullinates EP300/P300 at ′Arg-2142′, which favors its interaction with NCOA2/GRIP1.Specifications
| PADI4 | |
| Polyclonal | |
| Unconjugated | |
| PADI4 | |
| HL 60 PAD, PAD, PAD4, PADI4, PADI5, PDI4, PDI5, Peptidylarginine deiminase IV | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18602, 23569, 29512 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88807, Q9UM07, Q9Z183 | |
| Padi4 | |
| PADI4 Fusion Protein Ag11068 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction