Learn More
Kern Sohn Padded Wooden Box for Weights
Description
- Wooden
- For check weight design
Specifications
Specifications
| Product Type | Case |
| Classification | F1 |
| Case Material | Wood |
| For Use With (Equipment) | Check Weights |
| Holds | 10 kg |