Learn More
PACT Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28636-1-AP-150UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17531376
  • 4593.76 NOK / 150µL
Estimated Shipment: 03-05-2024
Description
The PACT antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human PACT. This antibody recognizes human, mouse, rat antigen. The PACT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PACT
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Western Blot
Unconjugated
150 μL
8575