Learn More
PACSIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13219-1-AP-20UL
 View more versions of this product
Product Code. 16861334
  • 1710.33 NOK / 20µL
Description
May play a role in vesicle formation and transport.
Specifications
Specifications
PACSIN1
Polyclonal
Unconjugated
PACSIN1
KIAA1379, PACSIN1, SDPI
Rabbit
Antigen Affinity Chromatography
RUO
23969, 29704, 29993
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BY11
Pacsin1
PACSIN1 Fusion Protein Ag4102
20 μL
Primary
Rat, Mouse, Human