missing translation for 'onlineSavingsMsg'
Learn More

Paclobutrazol, TRC

Product Code. 17904005
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904005 250 mg 250mg
17973115 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17904005 Supplier Toronto Research Chemicals Supplier No. TRCP133000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76738-62-0
Molecular Formula C15 H20 Cl N3 O
Molecular Weight (g/mol) 293.79
Synonym 1H-1,2,4-Triazole-1-ethanol, β-[(4-chlorophenyl)methyl]-α-(1,1-dimethylethyl)-, (αR,βR)-rel-, 1H-1,2,4-Triazole-1-ethanol, β-[(4-chlorophenyl)methyl]-α-(1,1-dimethylethyl)-, (R*,R*)-(±)-, rel-(αR,βR)-β-[(4-Chlorophenyl)methyl]-α-(1,1-dimethylethyl)-1H-1,2,4-triazole-1-ethanol, (2-RS,3RS)-1-(4-Chlorophenyl)-4,4-dimethyl-2-(1H-1,2,4-triazol-1-yl)pentan-3-ol, 1H-1,2,4-Triazole-1-ethanol, β-[(4-chlorophenyl)methyl]-α-(1,1-dimethylethyl)-, (R*,R*)-, AuStar, Bonzi, Bounty Flowable, Clipper, Clipper (triazole), Cultar, Duo Xiao Zuo, Friazole, ICI-PP 333, MET, Multi-effect triazole, PP 333, PP 333 (plant growth regulator), Paclobutrazol, Parlay, Smarect, Trimmit
IUPAC Name (2R,3R)-1-(4-chlorophenyl)-4,4-dimethyl-2-(1,2,4-triazol-1-yl)pentan-3-ol
SMILES CC(C)(C)[C@@H](O)[C@@H](Cc1ccc(Cl)cc1)n2cncn2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H20ClN3O/c1-15(2,3)14(20)13(19-10-17-9-18-19)8-11-4-6-12(16)7-5-11/h4-7,9-10,13-14,20H,8H2,1-3H3/t13-,14+/m1/s1
Formula Weight 293.1295
Chemical Name or Material Paclobutrazol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.