Learn More
Bryson Packaging™ Aluminum Foil
Product Code. 12379548
Change view
Click to view available options
Width (Metric):
100mm
500mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
12379548 500mm Each
12309558 100mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12379548 Supplier Bryson Packaging™ Supplier No. AKL300040J
  • ONLINE EXCLUSIVE PRICE
    630.27 NOK
    260.60 NOK / Pack of 1
    Save 369.67 NOK
    58% Off
Estimated Shipment: 19-11-2026
Suited for general wrapping purposes and many laboratory applications. Bryson Packaging™ Aluminum Foil is ideal for use in autoclaves. Available in 100mm wide, 18μM guage and 500mm wide, 12μM guage rolls. Each roll has 75m of quality packaging foil.
  • Suitable for general wrapping purposes and many laboratory applications, including:
    • sealing bottles
    • culture tubes
    • stopping leaks
    • reducing evaporation or heat losses
    • covering corks
    • making weighing boats or dishes
  • 75m rolls available in a variety of widths and gaugess
  • Ideal for use in autoclaves
  • Supplied in a cutter box
Specifications
Applications Autoclavable
Length (English) 246.1 ft.
Color Metallic Silver
Product Type Aluminum Foil in Cutter Box
Width (Metric) 500mm
Material Aluminum
Thickness 12 μm