Learn More
VACUUBRAND™ Package Rotary Vane Pump RZ 6 with FO/VS 16
GREENER_CHOICE
Product Code. 17879816
Change view
Click to view available options
Plug Type:
CEE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type unitSize
17879816 CEE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17879816 Supplier VACUUBRAND™ Supplier No. 20798039
Discount Lab Supplies
Use promo code 28170
View all promotions
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
The rotary vane pump package RZ 6 with oil mist filter and butterfly valve VS 16 offers great performance in a compact design - even with the gas ballast open, they enable a very good ultimate vacuum.
  • High flow rates even at vacuum levels approaching ultimate vacuum
  • High water vapor tolerance due to efficient gas ballast; very good ultimate vacuum even with gas ballast
  • Vacuum-tight at switch-off; external anti-suckback valve not needed
  • Large oil volume: Long intervals between oil changes
  • Ease of maintenance due to telescopic design
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 0.002 mbar
Plug Type CEE
Flow Rate 5.7 m³/hr.
Pump Type Rotary Vane Vacuum Pump
Enclosure Rating IP40
Applications Degassing, Lyophilization, Fine Vacuum Distillation and Drying Chambers
Maximum Pressure 1.1 bar
Voltage 230 V