Learn More
Fisherbrand™ Pack of Cryogenic and Vaccine Vial Gripper
Product Code. 17897145 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
2 Pk.
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17897145 2 Pk. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17897145
  • ONLINE EXCLUSIVE PRICE
    309.00 NOK
    267.07 NOK / Pack of 2
    Save 41.93 NOK
    13% Off
Estimated Shipment: 12-01-2027
The perfect grip for optimal precision.
  • Durably made of reinforced polypropylene which is cold-tolerant and resistant to everyday wear
  • The designated squeeze target ensures maximum precision and grip with every use
Specifications
Product Type Tongs
Quantity 2 Pk.
Sterility Non-sterile
Length (Metric) 15.24 cm
Material GFPP