missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PABPC1/PABP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10970-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a poly(A) binding protein. The protein shuttles between the nucleus and cytoplasm and binds to the 3′ poly(A) tail of eukaryotic messenger RNAs via RNA-recognition motifs. The binding of this protein to poly(A) promotes ribosome recruitment and translation initiation; it is also required for poly(A) shortening which is the first step in mRNA decay. The gene is part of a small gene family including three protein-coding genes and several pseudogenes.Specifications
| PABPC1/PABP | |
| Polyclonal | |
| Unconjugated | |
| PABPC1 | |
| PAB1, PABP, PABP 1, PABP1, PABPC1, PABPC2, PABPL1, Poly(A) binding protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171350, 18458, 26986 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P11940, P29341, Q9EPH8 | |
| PABPC1 | |
| PABPC1,PABP Fusion Protein Ag1422 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction