Learn More
PABPC1/PABP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16807713 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16807713 20 μL 20µL
16897703 150 μL 150µL
2 options
Product Code. 16807713
Brand: Proteintech 109701AP20UL
Rabbit Polyclonal Antibody
This gene encodes a poly(A) binding protein. The protein shuttles between the nucleus and cytoplasm and binds to the 3′ poly(A) tail of eukaryotic messenger RNAs via RNA-recognition motifs. The binding of this protein to poly(A) promotes ribosome recruitment and translation initiation; it is also required for poly(A) shortening which is the first step in mRNA decay. The gene is part of a small gene family including three protein-coding genes and several pseudogenes.
Specifications
Antigen PABPC1/PABP
Applications Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PABPC1
Gene Accession No. P11940, P29341, Q9EPH8
Gene Alias PAB1, PABP, PABP 1, PABP1, PABPC1, PABPC2, PABPL1, Poly(A) binding protein 1
Gene Symbols PABPC1
Host Species Rabbit
Immunogen PABPC1,PABP Fusion Protein Ag1422
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171350, 18458, 26986
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG