Learn More
PABPC1 PABP Mouse anti-Human, Unconjugated, Clone: 3D8B5, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | PABPC1 PABP |
|---|---|
| Clone | 3D8B5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16869025
|
Proteintech
66809-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16859025
|
Proteintech
66809-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PABPC1, PABP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PABPC1, PABP. This antibody recognizes human, rat antigen. The PABPC1, PABP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| PABPC1 PABP | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 26986 | |
| Primary |
| 3D8B5 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Rat | |
| IgG1 |