missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2RY12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11976-1-AP-20UL
This item is not returnable.
View return policy
Description
P2Y12 belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is involved in platelets aggregation, and is a potential target for the treatment of thromboembolisms and other clotting disorders. Two transcript variants encoding the same isoform have been identified for this gene. Expression of P2Y12 has been reported in platelets, spinal cord, and many brain regions. ESTs for P2Y12 have been isolated from B-cell/lung/testis, brain, embryo, prostate, eye, kidney carcinoma, and colon carcinoma libraries.Specifications
| P2RY12 | |
| Polyclonal | |
| Unconjugated | |
| P2RY12 | |
| ADP glucose receptor, ADPG R, HORK3, P2RY12, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12, P2Y12 platelet ADP receptor, SP1999 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64803, 64805, 70839 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9CPV9, Q9EPX4, Q9H244 | |
| P2ry12 | |
| P2RY12 Fusion Protein Ag2601 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction