Learn More
p-Terphenyl D14 in DCM 2000μg/mL, Fisher chemical™
Product Code. p-7083863 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Ampoule
Bottle
Unit Size:
1mL
5mL
Product Code. 12997103
Brand: Fisher Chemical P887010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
CAS 1718-51-0
Molecular Formula C18H14
Molecular Weight (g/mol) 244.395
InChI Key XJKSTNDFUHDPQJ-WZAAGXFHSA-N
PubChem CID 123128
IUPAC Name 1,2,3,4,5-pentadeuterio-6-[2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3
Specifications
Packaging Bottle
Analyte or Component Names p-Terphenyl D14
Concentration 2000 μg/mL
Type Pesticide and Metabolite Standards
Element p-Terphenyl D14
Solvent or Matrix Dichloromethane
Quantity 1 mL