missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenol, >99%, MP Biomedicals™

Product Code. p-4404285 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413730 1 g 1g
11423730 5 g 5g
11433730 25 g 25g
11443730 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11413730 Supplier MP Biomedicals Supplier No. 0210246101
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Color Yellow
Density 1.479g/mL
Quantity 1 g
Formula Weight 139.11
Percent Purity >99%
Physical Form Crystalline Powder
Chemical Name or Material p-Nitrophenol
compliance-icons compliance-icons
Product Identifier
  • p-Nitrophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H312+H332-Harmful in contact with skin or if inhaled
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.