missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenol, >99%, MP Biomedicals™

Product Code. 11423730 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11423730

Brand: MP Biomedicals 0210246105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Color Yellow
Density 1.479g/mL
Quantity 5 g
Formula Weight 139.11
Percent Purity >99%
Physical Form Crystalline Powder
Chemical Name or Material p-Nitrophenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.