Learn More
p-Iodonitrotetrazolium Violet, MP Biomedicals
Product Code. 11328909 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328909 100 mg 100mg
11338909 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11328909 Supplier MP Biomedicals Supplier No. 02102066.1
  • ONLINE EXCLUSIVE PRICE
    443.00 NOK
    421.09 NOK / 100mg
    Save 21.91 NOK
    4% Off
 Restricted Item
Estimated Shipment: 14-12-2026
  • p-Iodonitrotetazolium Violet is used as a electron acceptor for the colorimetric assay of various dehydrogenases
Chemical Identifiers
CAS 146-68-9
Molecular Formula C19H13ClIN5O2
Molecular Weight (g/mol) 505.70
MDL Number MFCD00011961,MFCD00149999
InChI Key JORABGDXCIBAFL-UHFFFAOYSA-M
Synonym iodonitrotetrazolium chloride, iodonitrotetrazolium, iodonitrotetrazolium purple, iodonitrotetrazolium violet, 2-4-iodophenyl-3-4-nitrophenyl-5-phenyltetrazolium chloride, iodonitro tetrazolium, int, p-iodonitrotetrazolium violet, int chemical, 2-p-iodophenyl-3-p-nitrophenyl-5-phenyltetrazolium chloride
PubChem CID 64957
ChEBI CHEBI:75421
IUPAC Name 3-(4-iodophenyl)-2-(4-nitrophenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
SMILES [Cl-].[O-][N+](=O)C1=CC=C(C=C1)[N+]1=NC(=NN1C1=CC=C(I)C=C1)C1=CC=CC=C1
Specifications
Melting Point 245°C to 251°C (literature)
Color Yellow
Linear Formula C19H13ClIN5O2
Quantity 100 mg
Solubility Information Soluble in 5% methanol/water(1:1)(clear yellow); slightly soluble in water, alcohol, DMSO.
Formula Weight 505.7
Physical Form Powder
Chemical Name or Material p-IODONITROTETRAZOLIUM VIOLET