missing translation for 'onlineSavingsMsg'
Learn More

p-Dimethylamino-Benzaldehyde, ACS, MP Biomedicals™

Product Code. 11402832 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
Product Code. Quantity unitSize
11402832 250 g 250g
11482822 50 g 50g
11492822 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11402832

Brand: MP Biomedicals 0215251783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Quantity 250 g
Formula Weight 149.2
Grade ACS Reagent
Physical Form Solid
Chemical Name or Material p-Dimethylamino-Benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.