missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidinium Chloride, Sigma-Aldrich™

Product Code. 15820751
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15820751 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15820751 Supplier Sigma Aldrich Supplier No. 8201030050
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 20265-97-8
Melting Point 217°C to 220°C
Molecular Formula 4-(CH3O)C6H4NH3Cl
MDL Number MFCD00036388
Quantity 50 g
Synonym p-Anisidinium chloride, p-Anisidinium chloride, 4-Methoxyanilinium chloride, p-Anisidinium chloride
Molecular Weight (g/mol) 159.61
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.